Learn More
Medchemexpress LLC Aztreonam (Standard) | 78110-38-0 | 99.6% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0129R100MG
Encompass_Preferred
Aztreonam (Standard) is an analytical reference standard. It is a synthetic monocyclic beta-lactam antibiotic exhibiting high affinity for penicillin-binding protein 3 (PBP-3), making it suitable for various research and analytical applications.
  • Analytical standard grade
  • Reference standard for assays
  • Suitable for qualitative research
  • Suitable for quantitative research
  • Suitable for methodological research
  • Commonly used in HPLC
  • Commonly used in GC
  • Commonly used in MS
Catalog No. 50-004-50866