Learn More
Medchemexpress LLC Aztreonam (Standard) | 78110-38-0 | 99.6% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0129R5MG
Encompass_Preferred
Aztreonam (Standard) is the analytical standard of Aztreonam, intended for research and analytical applications. This synthetic monocyclic beta-lactam antibiotic has a very high affinity for penicillin-binding protein 3 (PBP-3).
  • Reference standard supplied assay
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Suitable for HPLC
  • Suitable for GC
  • Suitable for MS
Catalog No. 50-004-50939