Learn More
Medchemexpress LLC Aztreonam (Standard) | 78110-38-0 | 99.6% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0129R50MG
Encompass_Preferred
Aztreonam (Standard) is an analytical standard intended for research and analytical applications. It is a synthetic monocyclic beta-lactam antibiotic, also known as SQ-26,776, which demonstrates a high affinity for penicillin-binding protein 3 (PBP-3). This product is suitable for qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Synthetic monocyclic beta-lactam antibiotic
  • High affinity for penicillin-binding protein 3 (PBP-3)
  • Suitable for HPLC, GC, and MS research experiments
Catalog No. 50-004-50766