Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azulene-1-carboxaldehyde 97.0+%, TCI America™
SDP

Catalog No. A27521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A27521G Supplier TCI America Supplier No. A27521G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7206-61-3
Molecular Formula C11H8O
Molecular Weight (g/mol) 156.184
MDL Number MFCD19301077
InChI Key CZRXLQPVJOJLML-UHFFFAOYSA-N
Synonym 1-Formylazulene
PubChem CID 11744870
IUPAC Name azulene-1-carbaldehyde
SMILES C1=CC=C2C=CC(=C2C=C1)C=O

Specifications

Color Purple-Red
Quantity 1 g
Formula Weight 156.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Azulene-1-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.