Learn More
Medchemexpress LLC Azulene | 275-51-4 | MFCD00003810 | 99.98% | C10H8 | 250 MG
SDP
Catalog No. 5000359182
Encompass_Preferred
Azulene (Cyclopentacycloheptene) is an isomer of naphthalene with high anti-HIV activity. It is isolated from the distillation of chamomile oil and serves as a scaffold in medicinal chemistry. Azulene is a bicyclic nonbenzenoid aromatic hydrocarbon characterized by a dipole moment due to its electron-rich five-membered ring and electron-deficient seven-membered ring.
  • Isomer of naphthalene with high anti-HIV activity.
  • Isolated from distillation of chamomile oil.
  • Serves as a scaffold in medicinal chemistry.
  • Bicyclic nonbenzenoid aromatic hydrocarbon.
  • Characterized by a dipole moment.
Catalog No. 50-003-59182 Supplier Medchemexpress LLC Supplier No. 250MGHYB0055
Azulene (Cyclopentacycloheptene) is an isomer of naphthalene with high anti-HIV activity. It is isolated from the distillation of chamomile oil and serves as a scaffold in medicinal chemistry. Azulene is a bicyclic nonbenzenoid aromatic hydrocarbon characterized by a dipole moment due to its electron-rich five-membered ring and electron-deficient seven-membered ring.
  • Isomer of naphthalene with high anti-HIV activity.
  • Isolated from distillation of chamomile oil.
  • Serves as a scaffold in medicinal chemistry.
  • Bicyclic nonbenzenoid aromatic hydrocarbon.
  • Characterized by a dipole moment.