Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azulene, 99%

Catalog No. p-61757
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC105181000 100 mg
AC105180010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105181000 Supplier Thermo Scientific Chemicals Supplier No. 105181000
Only null left
Add to Cart

CAS: 275-51-4 | C10H8 | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 275-51-4
Molecular Formula C10H8
Molecular Weight (g/mol) 128.17
MDL Number MFCD00003810
InChI Key CUFNKYGDVFVPHO-UHFFFAOYSA-N
Synonym cyclopentacycloheptene, azunamic, bicyclo 5.3.0 decapentaene, azulen, unii-82r6m9mglp, bicyclo 5.3.0-1,3,5,7,9-decapentaene, bicyclo 5.3.0-deca-2,4,6,8,10-pentaene, 82r6m9mglp, azulekeep
PubChem CID 9231
ChEBI CHEBI:31249
IUPAC Name azulene
SMILES C1=CC=C2C=CC=C2C=C1

Specifications

Melting Point 99.0°C to 101.0°C
Color Blue
Boiling Point 242.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 100 mg
Beilstein 05, II, 432
Merck Index 15, 919
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether and acetone
Formula Weight 128.17
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Azulene, 99%
Show More Show Less
EINECSNumber 205-993-6
RTECSNumber CO4570000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.