Learn More
Medchemexpress LLC Azulene (Cyclopentacycloheptene) | 275-51-4 | 99.97% | C10H8 | 100 MG
SDP
Catalog No. 5000432286
Encompass_Preferred
Azulene (Cyclopentacycloheptene) is an isomer of naphthalene with high anti-HIV activity. Isolated from the distillation of chamomile oil, it serves as a scaffold in medicinal chemistry.
  • Structural isomer of naphthalene
  • Bicyclic nonbenzenoid aromatic hydrocarbon
  • Exhibits high anti-HIV activity
  • Has a dipole moment due to electron-rich five-membered ring and electron-deficient seven-membered ring
Catalog No. 50-004-32286 Supplier Medchemexpress LLC Supplier No. HYB0055100MG
Azulene (Cyclopentacycloheptene) is an isomer of naphthalene with high anti-HIV activity. Isolated from the distillation of chamomile oil, it serves as a scaffold in medicinal chemistry.
  • Structural isomer of naphthalene
  • Bicyclic nonbenzenoid aromatic hydrocarbon
  • Exhibits high anti-HIV activity
  • Has a dipole moment due to electron-rich five-membered ring and electron-deficient seven-membered ring