Learn More
Cayman Chemical AzumolensodIum salt 10mg
SDP
Catalog No. 502969856
Encompass_Preferred
A muscle relaxant that inhibits the release of calcium from skeletal muscle sarcoplasmic reticulum; inhibits a component of store-operated calcium entry (SOCE) that is coupled to the skeletal muscle ryanodine receptor, with 20 µM azumolene causing a 70% reduction in SOCE in myotubes
Catalog No. 50-296-9856 Supplier Cayman Chemical Supplier No. 1646210 MG
A muscle relaxant that inhibits the release of calcium from skeletal muscle sarcoplasmic reticulum; inhibits a component of store-operated calcium entry (SOCE) that is coupled to the skeletal muscle ryanodine receptor, with 20 µM azumolene causing a 70% reduction in SOCE in myotubes