Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Azur B Certified 10 GR
SDP

Catalog No. 50980191
Encompass_Preferred

(Azur I; Methylene Azur B)
CAS #531-55-5 C₁₅H₁₆N₃SBr F.W. 350.29
Solubility: Water-soluble; Alcohol-slightly soluble

An important component of Giemsa's stain for blood protozoa. Used for staining semi-thin sections of plant tissue. Solubility: Water-soluble; Alcohol-slightly soluble. See Hoefert, L.L. (1968). Polychromatic stains for thin sections of Beta embedded in epoxy resin. Stain Tech. 43:145.

Catalog No. 50-980-191 Supplier Electron Microscopy Sciences Supplier No. 11200
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Azur I; Methylene Azur B)
CAS #531-55-5 C₁₅H₁₆N₃SBr F.W. 350.29
Solubility: Water-soluble; Alcohol-slightly soluble

An important component of Giemsa's stain for blood protozoa. Used for staining semi-thin sections of plant tissue. Solubility: Water-soluble; Alcohol-slightly soluble. See Hoefert, L.L. (1968). Polychromatic stains for thin sections of Beta embedded in epoxy resin. Stain Tech. 43:145.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.