Learn More
Electron Microscopy Sciences Azur II Certified 10 GR
SDP
Catalog No. 50980192
Encompass_Preferred
(Azur B/Methylene Blue 1:1)
F.W. 751.91
For the morphological details of marrow cells, nuclei, and bacteria.
Catalog No. 50-980-192 Supplier Electron Microscopy Sciences Supplier No. 11210
(Azur B/Methylene Blue 1:1)
F.W. 751.91
For the morphological details of marrow cells, nuclei, and bacteria.