Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azure A

Catalog No. p-7042096
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6134614 25 g
AAJ6134622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6134614 Supplier Thermo Scientific Chemicals Supplier No. J6134614
Only null left
Add to Cart

For staining cell granules and tissues | CAS: 531-53-3 | C14H14ClN3S | 291.797 g/mol

For staining cell granules and tissues

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-53-3
Molecular Formula C14H14ClN3S
Molecular Weight (g/mol) 291.797
MDL Number MFCD00012112
InChI Key NALREUIWICQLPS-UHFFFAOYSA-N
Synonym C.I. 52005
PubChem CID 13735
IUPAC Name (7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride
SMILES C[N+](=C1C=CC2=NC3=C(C=C(C=C3)N)SC2=C1)C.[Cl-]

Specifications

Melting Point 290°C (decomposition)
Beilstein 3922287
Merck Index 14,927
Formula Weight 291.8
Color Green
Quantity 25 g
Physical Form Powder
Chemical Name or Material Azure A
EINECSNumber 208-510-7
RTECSNumber SP5660000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.