Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azure A, Certified, Thermo Scientific™

Catalog No. AC401630100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC401630100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC401630100 Supplier Thermo Scientific Chemicals Supplier No. 401630100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-53-3
Molecular Formula C14H14ClN3S
Molecular Weight (g/mol) 291.797
MDL Number MFCD00012112
InChI Key NALREUIWICQLPS-UHFFFAOYSA-N
Synonym C.I. 52005
PubChem CID 13735
IUPAC Name (7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride
SMILES C[N+](=C1C=CC2=NC3=C(C=C(C=C3)N)SC2=C1)C.[Cl-]

Specifications

Absorption Ratio 0.94 to 1.2
Dye Content 55% min.
Beilstein 27, 392
Merck Index 15, 920
Infrared Spectrum Authentic
Lambda Max 620 to 634nm
Formula Weight 291.79
Grade Extra Pure
Color Green
Quantity 10 g
Physical Form Crystalline Powder
Chemical Name or Material Azure A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.