Learn More
Abcam Azure A chloride salt, blue nuclear staining component, 25G
Catalog No. 502704669
Encompass_Preferred
MW 291.8 g/mol. Important nuclear staining component of polychrome methylene blue. Used in Lillie's modified Nocht's method for paraffin sections and the NCCLS method for blood smears.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4669 Supplier Abcam Supplier No. AB14638225G
MW 291.8 g/mol. Important nuclear staining component of polychrome methylene blue. Used in Lillie's modified Nocht's method for paraffin sections and the NCCLS method for blood smears.
The product is subject to the following: Abcam Restricted Use Statement