Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azure A chloride, Sigma-Aldrich

Catalog No. 01259088 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-088 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-088 Supplier MilliporeSigma Supelco Supplier No. 86104925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The combination of azure A and silver staining is used for the quantification of glycosaminoglycans in agarose gels. It is also used for the staining of chromosomes in the nucleus. Azure A is a main component of the azure B/polychromed methylene blue-eosin stains family, also referred to as the Romanowsky stain. Azure A is a basic cationic dye.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-53-3
Molecular Weight (g/mol) 291.8 g/mol

Specifications

Color Blue/Green
Quantity 25 g
Percent Purity 70%
Grade Certified
Chemical Name or Material Azure A chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.