Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azure B Certified, MP Biomedicals™

Catalog No. 0219052005 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-190-52005 5 g
02-190-52001 1 g
ICN19052010 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-190-52005 Supplier MP Biomedicals, Inc Supplier No. 0219052005
Add to Cart
Add to Cart

Azure B Certified is used as a stain.

  • Azure B Certified is used for differentiating cellular RNA (purplish) and DNA (blue-green) in plant tissues.
  • Useful as a stain for Negri Bodies.
  • Can be used in the Romanowsky-type stains for blood and blood protozoa.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-55-5
Molecular Formula C15H16ClN3S

Specifications

InChI Key KFZNPGQYVZZSNV-UHFFFAOYSA-M
SMILES [Cl-].CNC1=CC=C2N=C3C=CC(=CC3=[S+]C2=C1)N(C)C
IUPAC Name 3-(dimethylamino)-7-(methylamino)-5λ⁴-phenothiazin-5-ylium chloride
Molecular Weight (g/mol) 305.82
Grade Certified
Color Index No. 52010
Quantity 5 g
Physical Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.