Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azure B Eosinate, Thermo Scientific™

Catalog No. AC190620250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC190620250 25 g
AC190621000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC190620250 Supplier Thermo Scientific Chemicals Supplier No. 190620250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62298-42-4
Molecular Formula C50H38Br4N6O5S2
Molecular Weight (g/mol) 1186.627
InChI Key NCBQBFXQRBYYGY-UHFFFAOYSA-N
PubChem CID 2724069
IUPAC Name dimethyl-[7-(methylamino)phenothiazin-3-ylidene]azanium;2',4',5',7'-tetrabromo-3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-diolate
SMILES CNC1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.CNC1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C(=C4OC5=C(C(=C(C=C35)Br)[O-])Br)Br)[O-])Br

Specifications

Quantity 25 g
Chemical Name or Material Azure B Eosinate
EINECSNumber 263-490-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.