Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azure B, MP Biomedicals™

Catalog No. ICN15041701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15041701 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15041701 Supplier MP Biomedicals, Inc Supplier No. 0215041701
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 531-55-5
Molecular Formula C15H16ClN3S
Molecular Weight (g/mol) 305.824
MDL Number MFCD00011935
InChI Key DNDJEIWCTMMZBX-UHFFFAOYSA-N
Synonym C.I. 52010, N, N, N-Trimethylthionin
PubChem CID 68275
IUPAC Name dimethyl-[7-(methylamino)phenothiazin-3-ylidene]azanium;chloride
SMILES CNC1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-]

Specifications

Color Brown
Quantity 1 g
Physical Form Powder
Formula Weight 305.83
Chemical Name or Material Azure B, Stains and dyes
EINECSNumber 208-511-2
Recommended Storage Store at Room Temperature (15° to 30°C)
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.