Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azure B, Thermo Scientific™

Catalog No. AC401640100
Change view
Click to view available options
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC401640100 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC401640100 Supplier Thermo Scientific Chemicals Supplier No. 401640100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 531-55-5
Absorption Ratio 1.02 to 1.27
Dye Content 85% min.
Molecular Formula C15H16ClN3S
MDL Number MFCD00011935
Merck Index 15, 921
Synonym Azure I, C.I. 52010
InChI Key DNDJEIWCTMMZBX-UHFFFAOYSA-N
SMILES CNC1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-]
Infrared Spectrum Authentic
IUPAC Name dimethyl-[7-(methylamino)phenothiazin-3-ylidene]azanium;chloride
Molecular Weight (g/mol) 305.824
PubChem CID 68275
Lambda Max 640 to 655nm
Formula Weight 305.82
Grade Extra Pure
Color Green
Quantity 10g
Chemical Name or Material Azure B, Pure
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.