Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azure II Eosinate, Sigma-Aldrich

Catalog No. 01258985 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-985 100 g
01-258-862 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-985 Supplier MilliporeSigma Supelco Supplier No. 200220100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chief ingredient of Giemsa Stain.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53092-85-6

Specifications

Form Powder
Quantity 100 g
Product Type Methylene Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.