Learn More
Sino Biological Azurocidin / CAP37 / AZU1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10660T08100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Azurocidin / CAP37 / AZU1 (rh Azurocidin / CAP37 / AZU1; Catalog#10660-H08H; NP_001691.1; Met1-Pro250). Azurocidin / CAP37 / AZU1 specific IgG was purified by Human Azurocidin / CAP37 / AZU1 affinity chromatography.
Catalog No. 50-224-6332