Learn More
ACROBiosystems Recombinant Protein;Human Azurocidin / CAP37 / AZU1 Protein;HEK293;100UG;AZ1,AZU1,Azurocidin,HBP,AZAMP,AZU,CAP37,HUMAZUR,NAZC
Catalog No. 501659043
Encompass_Preferred
Recombinant Protein;Human Azurocidin / CAP37 / AZU1 Protein;100UG;Human AZU1, His Tag (AZ1-H5225) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Pro 250 (Accession # AAH69495).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.0 kDa. The protein migrates as 34-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9043 Supplier ACROBiosystems Supplier No. AZ1H5225100UG
Recombinant Protein;Human Azurocidin / CAP37 / AZU1 Protein;100UG;Human AZU1, His Tag (AZ1-H5225) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Pro 250 (Accession # AAH69495).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.0 kDa. The protein migrates as 34-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.