Learn More
Selleck Chemical LLC Azvudine-E1243-5MG
SDP
Supplier:  Selleck Chemical LLC E12435MG
Encompass_Preferred
Azvudine (RO-0622) is a novel nucleoside reverse transcriptase inhibitor with antiviral activity against human immunodeficiency virus (HIV) hepatitis B virus (HBV) and hepatitis C virus (HCV) potently inhibits HIV-1 (EC50 range 0 03 to 6 92 nM) and HIV-2 (EC50s ranging from 0 018 to 0 025 nM)
Catalog No. 50-239-0020