Learn More
Azzota Corp Stirring Heating Mantle, Capacity: 250Ml , Maximum Temperature Is 716'F (380'C). Stir Speed: 0 To 1400Rpm
Catalog No. NC1089133
Encompass_Preferred
Stirring Heating Mantle, Capacity: 250ml , Maximum temperature is 716'F (380'C). Stir speed: 0 to 1400rpm
Catalog No. NC1089133 Supplier Azzota Corp Supplier No. SHM250
Stirring Heating Mantle, Capacity: 250ml , Maximum temperature is 716'F (380'C). Stir speed: 0 to 1400rpm