Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ B-27™ Supplement (50X), custom

Catalog No. 0080085SA
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
00-800-85SA 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 00-800-85SA Supplier Gibco™ Supplier No. 0080085SA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The custom B-27 Supplement (50X) is a serum-free additive for the growth and expansion of retinal ganglion cells.

The custom B-27 Supplement (50X) is a serum-free additive for the growth and expansion of retinal ganglion cells. It is supplied as a 50X liquid concentrate and it is used with a basal medium, such as Neurobasal, and other additives, such as GlutaMAX-1.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10 mL - 50X frozen concentrate; store at -5°C to -20°C in the dark.
Cell Type Neural Cells
Classification Serum-free
Concentration 50 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Serum Level Serum-Free
Sterility Sterile
With Additives Antioxidant Cocktail, Insulin, Vitamin A
Endotoxin Level ≤ 25 EU/ml
pH 6 to 8
Product Line Gibco
Purity or Quality Grade Cell Culture Grade
Quantity 10 mL
Shipping Condition Dry Ice
Source Serum-Free
Tested For Sterility Testing (Bacteria & Fungi), Endotoxin (EU/ml), Growth Performance Assay
Show More Show Less
Do you offer B-27 supplements with and without insulin?

Yes, we do offer B-27 Supplement, minus insulin (Cat. Nos. A1895601 and A1895602). All our other B-27 supplements do contain insulin.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.