Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Beta-Actin Monoclonal Antibody, Mouse, HRP Conjugate, 100 μl
SDP

Catalog No. NC0977044
Encompass

This antibody is a HRP-conjugated mouse monoclonal antibody specifically directed against endogenous levels of total β-actin. The conjugate is formed by linking horseradish peroxidase (HRP) to the amine groups of the antibody, maintaining the species cross-reactivity profile of the parent clone 8H10D10. It is suitable for detecting β-actin, a ubiquitous cytoskeletal component.

  • Host species: mouse
  • Isotype: IgG2b
  • Validated application: western blotting
  • Species reactivity: human, mouse, rat, hamster, monkey, and dog
  • Formulation: 140 mM NaCl, 3 mM KCl, 10 mM sodium phosphate (pH 7.4), 2 mM potassium phosphate monobasic, 2 mg/mL BSA, and 50% glycerol
  • Storage conditions: −20°C (do not aliquot)

Catalog No. NC0977044 Supplier Cell Signaling Technology Supplier No. 12262S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This antibody is a HRP-conjugated mouse monoclonal antibody specifically directed against endogenous levels of total β-actin. The conjugate is formed by linking horseradish peroxidase (HRP) to the amine groups of the antibody, maintaining the species cross-reactivity profile of the parent clone 8H10D10. It is suitable for detecting β-actin, a ubiquitous cytoskeletal component.

  • Host species: mouse
  • Isotype: IgG2b
  • Validated application: western blotting
  • Species reactivity: human, mouse, rat, hamster, monkey, and dog
  • Formulation: 140 mM NaCl, 3 mM KCl, 10 mM sodium phosphate (pH 7.4), 2 mM potassium phosphate monobasic, 2 mg/mL BSA, and 50% glycerol
  • Storage conditions: −20°C (do not aliquot)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.