Learn More
Lonza Walkersville B-ALI SingleQuots Kit
Catalog No. 5000496219
Encompass
Culture system containing B-ALI Basal Media B-ALI Differentiation Media and B-ALI SingleQuots Supplements required for air-liquid interface differentiation of bronchial epithelial cells
Catalog No. 50-004-96219 Supplier Lonza Walkersville Supplier No. 00193515
Culture system containing B-ALI Basal Media B-ALI Differentiation Media and B-ALI SingleQuots Supplements required for air-liquid interface differentiation of bronchial epithelial cells