Learn More
Medchemexpress LLC B-AMINOPROPIONITRILE,500MG
SDP
Catalog No. NC1840357
Encompass_Preferred
β-Aminopropionitrile is a specific and irreversible lysyl oxidase (LOX) inhibitor. β-Aminopropionitrile targets the active site of LOX or LOXL isoenzymes.
Catalog No. NC1840357 Supplier Medchemexpress LLC Supplier No. HYY1750500MG
β-Aminopropionitrile is a specific and irreversible lysyl oxidase (LOX) inhibitor. β-Aminopropionitrile targets the active site of LOX or LOXL isoenzymes.