Learn More
Enzo Life Sciences beta-Amyloid (1-40) monoclonal antibody (11A50-B10) (1ml)
SDP
Catalog No. 502012673
Encompass_Preferred
Clone: 11A50-B10. Isotype: IgG1. Species reactivity: Human, Mouse. Specificity: Recognizes C-terminal β-amyloid. Specific for the isoform ending at aa40. Applications: ELISA, IHC, WB. Purity: Protein G purified. Formulation: Liquid. In PBS. Does not contain any preservatives or carrier proteins.
Catalog No. 50-201-2673 Supplier Enzo Life Sciences Supplier No. ENZABS1161000
Clone: 11A50-B10. Isotype: IgG1. Species reactivity: Human, Mouse. Specificity: Recognizes C-terminal β-amyloid. Specific for the isoform ending at aa40. Applications: ELISA, IHC, WB. Purity: Protein G purified. Formulation: Liquid. In PBS. Does not contain any preservatives or carrier proteins.