Learn More
Enzo Life Sciences beta-Amyloid (1-40) monoclonal antibody (11A50-B10) (200µl)
SDP
Supplier:  Enzo Life Sciences ENZABS1160200
Encompass_Preferred
Clone: 11A50-B10. Isotype: IgG1. Species reactivity: Human, Mouse. Specificity: Recognizes C-terminal β-amyloid. Specific for the isoform ending at aa40. Applications: ELISA, IHC, WB. Purity: Protein G purified. Formulation: Liquid. In PBS. Does not contain any preservatives or carrier proteins.
Catalog No. 50-201-2672