Learn More
Cusabio Technology LLC B and T lymphocyte associated
Catalog No. 5000048905
Encompass_Preferred
This kit detects BTLA in mouse serum plasma and tissue homogenates using a quantitative sandwich ELISA method It has a detection range of 0 625 ng/mL to 40 ng/mL and a sensitivity of 0 156 ng/mL ensuring accurate measurement of low-abundance targets The assay requires 50-100 L of sample and can be completed within 1-5 hours Results are determined by measuring absorbance at 450 nm providing a reliable tool for researchers studying BTLA s role in immune cell regulation lymphocyte attenuation or related immunological pathways The standardized protocol supports reproducibility across experimental conditions
Catalog No. 50-000-48905 Supplier Cusabio Technology LLC Supplier No. CSBEL002868MO
This kit detects BTLA in mouse serum plasma and tissue homogenates using a quantitative sandwich ELISA method It has a detection range of 0 625 ng/mL to 40 ng/mL and a sensitivity of 0 156 ng/mL ensuring accurate measurement of low-abundance targets The assay requires 50-100 L of sample and can be completed within 1-5 hours Results are determined by measuring absorbance at 450 nm providing a reliable tool for researchers studying BTLA s role in immune cell regulation lymphocyte attenuation or related immunological pathways The standardized protocol supports reproducibility across experimental conditions