Learn More
Medchemexpress LLC B-AP15 | 1009817-63-3 | 98.1% | 419.39 | 200 MG
SDP
Catalog No. 5000348867
Encompass_Preferred
B-AP15 is a specific inhibitor of the deubiquitinating enzymes UCHL5 and Usp14. It is for research use only.
  • Inhibits UCHL5 and Usp14 deubiquitinating enzymes
  • Induces tumor cell apoptosis
  • Available as a solid, light yellow to yellow appearance
  • Formula: C22H17N3O6
Catalog No. 50-003-48867 Supplier Medchemexpress LLC Supplier No. HY13989200MG
B-AP15 is a specific inhibitor of the deubiquitinating enzymes UCHL5 and Usp14. It is for research use only.
  • Inhibits UCHL5 and Usp14 deubiquitinating enzymes
  • Induces tumor cell apoptosis
  • Available as a solid, light yellow to yellow appearance
  • Formula: C22H17N3O6