Learn More
Medchemexpress LLC HY-13989 10mg Medchemexpress, b-AP15 CAS:1009817-63-3 Purity:>98%
SDP
Catalog No. 502029042
Encompass_Preferred
Medchemexpress, HY-13989 10mg b-AP15 CAS:1009817-63-3 b-AP15 is a specific inhibitor of the deubiquitinating enzymes UCHL5 and Usp14. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9042 Supplier Medchemexpress LLC Supplier No. HY1398910MG
Add to Cart
Add to Cart
Medchemexpress, HY-13989 10mg b-AP15 CAS:1009817-63-3 b-AP15 is a specific inhibitor of the deubiquitinating enzymes UCHL5 and Usp14. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.