Learn More
Bachem (b-Asp5)-Delta-Sleep Inducing Peptide Trifluoroacetate
SDP
Supplier:  Bachem 4028635.0005
Encompass_Preferred
(b-Asp5)-Delta-Sleep Inducing Peptide Trifluoroacetate, H-Trp-Ala-Gly-Gly-Asp(Ala-Ser-Gly-Glu-OH)-OH, 82602-88-8, C35H48N10O15, Mr 848.82, 5mg. (b-Asp5)-Delta-Sleep Inducing Peptide (rabbit), (b-Asp5)-DSIP (rabbit). This DSIP analog serves as a good substrate for protein L-isoaspartyl methyltransferase, a n enzyme occuring in the brain involved in the repair of age-damaged proteins containing atypical iosaspartyl peptide bonds.
Catalog No. 50-259-621