Learn More
Hanna Pharmaceutical Supply Co., Inc. IV EXTENSION SET 10" 50/CS
Catalog No. NC1763188
Encompass_Preferred
5.0 Micron Air-eliminating In-line Filter with Female Luer Connector, Male SPIN-LOCK Connector. DEHP-free, Latex-free. Priming Volume: 3.6 mL. Length: 10 in. (25.4 cm). 50/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
This item is not returnable. View return policy
Catalog No. NC1763188 Supplier Hanna Pharmaceutical Supply Co., Inc. Supplier No. BBR474002
5.0 Micron Air-eliminating In-line Filter with Female Luer Connector, Male SPIN-LOCK Connector. DEHP-free, Latex-free. Priming Volume: 3.6 mL. Length: 10 in. (25.4 cm). 50/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.