Learn More
Anton Paar 5 ml Luer Syringes (100 pcs)
Catalog No. NC0423474
Encompass_Preferred
Pack of 100 Luer Tip Syringes (5 ml) used to fill sample into the measuring cell of DMA density meters.Suitable for use with Anton Paar density metersDMA 501 Compact density and concentration meter NC1818227,DMA 1001 Precision compact density and concentration meter NC1818230
Catalog No. NC0423474 Supplier Anton Paar Supplier No. 6772
Pack of 100 Luer Tip Syringes (5 ml) used to fill sample into the measuring cell of DMA density meters.Suitable for use with Anton Paar density metersDMA 501 Compact density and concentration meter NC1818227,DMA 1001 Precision compact density and concentration meter NC1818230