Learn More
Seracare Life Sciences Inc B BURGDORFERI POS CONTROL
Catalog No. 5067469
Encompass_Preferred
Heat-killed Borrelia burgdorferi cells. It consists of at least 1 mL of approximately 1% w/v wet packed cells in dextran solution. It is intended for use as a positive control in immunoassays designed for Borrelia detection.
This item is not returnable. View return policy
Catalog No. 50-674-69 Supplier Seracare Life Sciences Inc Supplier No. 53700023
Heat-killed Borrelia burgdorferi cells. It consists of at least 1 mL of approximately 1% w/v wet packed cells in dextran solution. It is intended for use as a positive control in immunoassays designed for Borrelia detection.