Learn More
OPTI MEDICAL B-CASSETTES
Catalog No. NC9053836
Encompass
25 Cassettes per box, BP7562, pH, PO2, PCO2, tHb, SO2; For OPTI CCa analyzer
Catalog No. NC9053836 Supplier OPTI MEDICAL Supplier No. BP7562
25 Cassettes per box, BP7562, pH, PO2, PCO2, tHb, SO2; For OPTI CCa analyzer