Learn More
Enzo Life Sciences beta-Catenin (NT/exon 2) monoclonal antibody (7D11) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804060C100
Encompass_Preferred
Clone: 7D11. Isotype: IgG2a. Immunogen: Recombinant human β-catenin. Species reactivity: Human, Mouse, Dog. Specificity: Recognizes the N-terminus (exon 2) of human, mouse, rat (weak) and dog β-catenin. Applications: ELISA, IHC, IP, WB. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-9764