Learn More
ECM Bioscience β-Catenin Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience CK6120
Encompass_Preferred
The β-Catenin phospho-regulation antibody sampler kit can be used to examine phosphorylation of β-Catenin at Tyr-86, Tyr-142, Tyr-489 and Tyr-654. The kit also includes monoclonal and polyclonal antibodies to monitor total expression levels of β-Catenin. Applications: WB, E, ICC, IHC, IP; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8622