Learn More
ALADDIN SCIENTIFIC CORPORATION B-Cyclodextrin, 100g
Catalog No. 5000082551
Encompass_Preferred
Useful for forming water-soluble complexes for fluorescent labeling of proteins Product Application-Use to solubilize non-polar compounds such as fatty acids lipids and cholesterol Reported useful for the selective precipitation of enantiomeric positional or structural isomers-Cyclodextrin is used with dansyl chloride to form water-soluble complexes for fluorescent labeling of proteins It is an active ingredient of household odor eliminator It is also used in personal care products like toothpastes skin creams and dusting powders It finds applications in the cosmetic industry for products like detergents and perfumes for the controlled release of fragrances Further it is used to produce HPLC columns allowing chiral enantiomers separation In addition to this it is used to decrease the level of cholesterol in milk fat
Catalog No. 50-000-82551 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. C104384100G
Useful for forming water-soluble complexes for fluorescent labeling of proteins Product Application-Use to solubilize non-polar compounds such as fatty acids lipids and cholesterol Reported useful for the selective precipitation of enantiomeric positional or structural isomers-Cyclodextrin is used with dansyl chloride to form water-soluble complexes for fluorescent labeling of proteins It is an active ingredient of household odor eliminator It is also used in personal care products like toothpastes skin creams and dusting powders It finds applications in the cosmetic industry for products like detergents and perfumes for the controlled release of fragrances Further it is used to produce HPLC columns allowing chiral enantiomers separation In addition to this it is used to decrease the level of cholesterol in milk fat