Learn More
Medchemexpress LLC B-Cyclodextrin 1kg | 7585-39-9 | 1KG
SDP
Catalog No. 5000240428
Encompass_Preferred
-Cyclodextrin is a cyclic polysaccharide composed of seven units of glucose ( -D-glucopyranose) linked by -(1 4) type bonds -Cyclodextrin has often been used to enhance the solubility of agents -Cyclodextrin has anti-influenza virus H1N1 activities
Catalog No. 50-002-40428 Supplier Medchemexpress LLC Supplier No. HY1072011KG
-Cyclodextrin is a cyclic polysaccharide composed of seven units of glucose ( -D-glucopyranose) linked by -(1 4) type bonds -Cyclodextrin has often been used to enhance the solubility of agents -Cyclodextrin has anti-influenza virus H1N1 activities