Learn More
Cole-Parmer BD Biocoat Disposable Syringe, Non-Sterile, Luer-Lok, 10 mL, 50/Pk
Catalog No. NC1096042 Shop All Cole-Parmer Products
Encompass_Preferred
Use these latex-free, disposable polypropylene syringes for your general-purpose applications. Available in either Luer-Lok or slip-tip versions. Syringes come nonsterilized in bulk packages and can be autoclaved. Needles and shield tips are not included; order separately.
Catalog No. NC1096042 Supplier Cole-Parmer Supplier No. 0794412
Use these latex-free, disposable polypropylene syringes for your general-purpose applications. Available in either Luer-Lok or slip-tip versions. Syringes come nonsterilized in bulk packages and can be autoclaved. Needles and shield tips are not included; order separately.