Learn More
Bachem B-Endorphin (human) Trifluoroacetate
SDP
Supplier:  Bachem 4006426.0005
Encompass_Preferred
B-Endorphin (human) Trifluoroacetate, H-Tyr-Gly-Gly-Phe-Met-Thr-Ser-Glu-Lys-Ser-Gln-Thr-Pro-Leu-Val-Thr-Leu-Phe-Lys-Asn-Ala-Ile-Ile-Lys-Asn-Ala-Tyr-Lys-Lys-Gly-Glu-OH, 61214-51-5, C158H251N39O46S, Mr 3465.03, 5mg. b-Lipotropin (59-89) (human), Lipotropin C Fragment (human).
Catalog No. 50-259-643