Learn More
Santa Cruz Biotechnology β-Estradiol 3-sulfate sodium salt, 25 mg | CAS# 4999-79-5
Catalog No. NC3343484
Encompass
β-Estradiol 3-sulfate sodium salt functions as a steroid hormone and exhibits estrogenic activity. It acts by binding to estrogen receptors, estrogen receptor alpha and beta, within the cell nucleus. This binding initiates a cascade of molecular events, leading to the regulation of gene expression and the modulation of various cellular processes. β-Estradiol 3-sulfate sodium salt′s mechanism of action involves influencing the transcription of target genes, thereby impacting the physiological responses of the cells. By interacting with estrogen receptors, β-Estradiol 3-sulfate sodium salt plays a role in the regulation of cell proliferation, differentiation, and apoptosis. Its activity at the molecular level contributes to the modulation of various signaling pathways and cellular functions, making it a relevant component in studies exploring hormone signaling and related biological processes.
Catalog No. NC3343484 Supplier Santa Cruz Biotechnology Supplier No. SC239863
β-Estradiol 3-sulfate sodium salt functions as a steroid hormone and exhibits estrogenic activity. It acts by binding to estrogen receptors, estrogen receptor alpha and beta, within the cell nucleus. This binding initiates a cascade of molecular events, leading to the regulation of gene expression and the modulation of various cellular processes. β-Estradiol 3-sulfate sodium salt′s mechanism of action involves influencing the transcription of target genes, thereby impacting the physiological responses of the cells. By interacting with estrogen receptors, β-Estradiol 3-sulfate sodium salt plays a role in the regulation of cell proliferation, differentiation, and apoptosis. Its activity at the molecular level contributes to the modulation of various signaling pathways and cellular functions, making it a relevant component in studies exploring hormone signaling and related biological processes.