Learn More
Sigma Aldrich Fine Chemicals Biosciences β-Glucuronidase from Escherichia coli | CAS 9027-70-9 | 1,000,000 Units
Catalog No. NC2480242
Encompass
β-glucuronidase from E. coli | Purity: ≥90% | Mol Wt: 68 kDa | CAS No: 9027-70-9 | Unit: 1,000,000 Units
Catalog No. NC2480242 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G0251500KU
β-glucuronidase from E. coli | Purity: ≥90% | Mol Wt: 68 kDa | CAS No: 9027-70-9 | Unit: 1,000,000 Units