Learn More
Apexbio Technology LLC B-HT-920-DIHYDROCHLORIDE-100MG
SDP
Catalog No. 5000569444
Encompass_Preferred
B-HT 920 dihydrochloride (CAS No 36085-73-1) is a research chemical frequently used in the study of neurological pathways due to its role as an agonist for dopamine D2 receptors It originates from biochemical research dedicated to understanding neuromodulation in both physiological and pathological states Its mechanism involves activating the D2 receptor influencing dopaminergic signaling pathways which play a crucial role in motor control reward and endocrine regulation In vitro B-HT 920 dihydrochloride exhibits activity in the nanomolar to low micromolar range making it effective for precise studies in cell models and receptor binding assays It is commonly utilized in cell-based experiments animal studies focused on Parkinson s disease models and drug screening processes Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-69444 Supplier Apexbio Technology LLC Supplier No. A3239100
B-HT 920 dihydrochloride (CAS No 36085-73-1) is a research chemical frequently used in the study of neurological pathways due to its role as an agonist for dopamine D2 receptors It originates from biochemical research dedicated to understanding neuromodulation in both physiological and pathological states Its mechanism involves activating the D2 receptor influencing dopaminergic signaling pathways which play a crucial role in motor control reward and endocrine regulation In vitro B-HT 920 dihydrochloride exhibits activity in the nanomolar to low micromolar range making it effective for precise studies in cell models and receptor binding assays It is commonly utilized in cell-based experiments animal studies focused on Parkinson s disease models and drug screening processes Experimental concentrations typically depend on the specific research design