Learn More
Cayman Chemical B-HT 920 (hydrochloride), Purity: ≥95%, CAS: 36085-73-1, 10 mg
SDP
Catalog No. 502953633
Encompass_Preferred
A potent agonist of D2 receptors (Ki = 5.8 nM); has neuroprotective and regenerative effects in animal models of Parkinson’s disease
Catalog No. 50-295-3633 Supplier Cayman Chemical Supplier No. 1417710 MG
A potent agonist of D2 receptors (Ki = 5.8 nM); has neuroprotective and regenerative effects in animal models of Parkinson’s disease