Learn More
Medchemexpress LLC B I09 | 1607803-67-7 | 99.4% | 303.31 | C16H17NO5 | 50 MG
SDP
Catalog No. 5000239816
Encompass_Preferred
B I09 is a small-molecule research reagent that inhibits the IRE-1 RNase. It is intended for in vitro studies of endoplasmic reticulum stress and related signaling pathways; solutions are unstable and should be freshly prepared prior to use.
  • Inhibits IRE-1 RNase with an IC50 of 1,230 nM.
  • Suitable for in vitro research of ER stress and XBP1-related pathways.
  • High purity (99.4%) supports reproducible experimental results.
  • Supplied as a solid with a molecular weight of 303.31 for accurate dosing.
  • Store at 4°C and protect from light; prepare solutions fresh due to instability.
  • Available in multiple package sizes to accommodate small-scale and larger assays.
Catalog No. 50-002-39816 Supplier Medchemexpress LLC Supplier No. HY10740050MG
B I09 is a small-molecule research reagent that inhibits the IRE-1 RNase. It is intended for in vitro studies of endoplasmic reticulum stress and related signaling pathways; solutions are unstable and should be freshly prepared prior to use.
  • Inhibits IRE-1 RNase with an IC50 of 1,230 nM.
  • Suitable for in vitro research of ER stress and XBP1-related pathways.
  • High purity (99.4%) supports reproducible experimental results.
  • Supplied as a solid with a molecular weight of 303.31 for accurate dosing.
  • Store at 4°C and protect from light; prepare solutions fresh due to instability.
  • Available in multiple package sizes to accommodate small-scale and larger assays.