Learn More
Medchemexpress LLC B I09 | 1607803-67-7 | 99.4% | 303.31 g/mol | C16H17NO5 | 100 MG
SDP
Catalog No. 5000239992
Encompass_Preferred
B I09 is a small-molecule inhibitor of the IRE-1 RNase with a reported in vitro IC50 of 1,230 nM. It inhibits splicing of XBP1 mRNA and reduces expression of XBP-1s in cellular assays, and is used to probe IRE-1-mediated unfolded protein response signaling. The compound is a light yellow to brown solid, soluble in DMSO, and unstable in solution so solutions should be freshly prepared.
Catalog No. 50-002-39992 Supplier Medchemexpress LLC Supplier No. HY107400100MG
Add to Cart
Add to Cart
B I09 is a small-molecule inhibitor of the IRE-1 RNase with a reported in vitro IC50 of 1,230 nM. It inhibits splicing of XBP1 mRNA and reduces expression of XBP-1s in cellular assays, and is used to probe IRE-1-mediated unfolded protein response signaling. The compound is a light yellow to brown solid, soluble in DMSO, and unstable in solution so solutions should be freshly prepared.