Learn More
Electron Microscopy Sciences Classic Eyeglass Single Lens Loupe 3x
SDP
Catalog No. 5024597
Encompass_Preferred
The eyeglass loupes are ideal for any detail work and are made from precision optical glass. Features improved mounting system for more secure attachment. High quality case provided to protect your investment. Lens diameter is 24.0 mm.
Catalog No. 50-245-97 Supplier Electron Microscopy Sciences Supplier No. 6825035
The eyeglass loupes are ideal for any detail work and are made from precision optical glass. Features improved mounting system for more secure attachment. High quality case provided to protect your investment. Lens diameter is 24.0 mm.